Learn More
Chicken anti-Rat IgG (H+L) Secondary Antibody, DyLight 680, Novus Biologicals™
Chicken Polyclonal Antibody
Brand:  Novus Biologicals NBP1-75363FR
Product Code. 18434085
  • € 389.00 / 1mg
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
Western Blot : 1:100-1:2000, ELISA : 1:100-1:2000, Immunohistochemistry : 1:10 - 1:500, Immunoprecipitation : 1:10 - 1:500, Immunodiffusion, Immunoassay
Chicken
Affinity purified
RUO
Secondary
Store at 4°C in the dark.
IgG
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion
DyLight 680
50mM Sodium Borate
This Chicken anti-Rat IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against purified rat IgG (H&L).
1 mL
Adaptive Immunity, Immunology
Rat