Learn More
Chicken anti-Rat IgG whole molecule, HRP, Polyclonal, Secondary Antibody, Abnova™
Brand:  Abnova PAB10846.1mg
Product Code. 16213140
  • € 403.00 / 1mg
Specifications
Chicken anti-Rat IgG whole molecule
Polyclonal
Chicken anti-rat IgG recognizes rat IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase.
Lyophilized from 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 (0.01% (w/v) gentamicin sulfate (Do NOT add sodium azide!), 10mg/mL BSA (Immunoglobulin and Protease free))
Rat IgG whole molecule
1 mg
Rat
Lyophilized
Dot Blot, ELISA, Immunohistochemistry, Western Blot
HRP
The optimal working dilution should be determined by the end user.
Chicken
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Secondary
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.