Learn More
SDHAF2, Chicken, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against synthetic peptide of SDHAF2.
Brand: Abnova PAB25763.100ug
Description
The SDHAF2 gene encodes a protein necessary for the flavination of the succinate dehydrogenase complex subunit SDHA (MIM 600857) and thus for SDH activity (Hao et al., 2009 [PubMed 19628817]).[supplied by OMIM
Specifications
| SDHAF2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| SDHAF2 | |
| SDHAF2 | |
| A synthetic peptide corresponding to 16 amino acids at N-terminus of human SDHAF2. | |
| 100 μg | |
| Primary | |
| SDHAF2 antibody is predicted to not cross-react with other SDHAF protein family members. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgY |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Chicken polyclonal antibody raised against synthetic peptide of SDHAF2. | |
| In PBS (0.02% sodium azide) | |
| C11orf79/PGL2/SDH5 | |
| Chicken | |
| Peptide affinity purification | |
| RUO | |
| 54949 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only