missing translation for 'onlineSavingsMsg'
Learn More

CHID1 Antibody (CHID1/6368), Novus Biologicals™

Mouse Monoclonal Antibody

€ 232.00 - € 462.00

Specifications

Antigen CHID1
Clone CHID1/6368
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18640955
View Documents
Novus Biologicals
NBP3-23888-100ug
100 μg
€ 462.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18684964
View Documents
Novus Biologicals
NBP3-23888-20ug
20 μg
€ 232.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CHID1 Monoclonal antibody specifically detects CHID1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

CHID1
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Lipid and Metabolism
10 mM PBS with 0.05% BSA
66005
IgG1 κ
Protein A or G purified
CHID1/6368
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
chitinase domain containing 1, chitinase domain-containing protein 1, FLJ42707, MGC3234, SI-CLP, stabilin-1 interacting chitinase-like protein, Stabilin-1-interacting chitinase-like protein
Recombinant full-length human CHID1 protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.