missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chikungunya Virus E1 Recombinant Rabbit Monoclonal Antibody (HL2069)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA547277

Product Code. 17849335

  • € 474.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Chikungunya fever is a pandemic disease caused by the mosquito-borne Chikungunya virus (CHIKV). The virus E1 glycoprotein mediation of viral membrane fusion during CHIKV infection is a crucial step in the release of viral genome into the host cytoplasm for replication.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chikungunya Virus E1
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Rabbit
Protein A
RUO
Virus
Antibody
IgG
Western Blot
HL2069
Unconjugated
Chikungunya Virus
Recombinant fragment of Chikungunya virus E1.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.