missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Chikungunya Virus E3 Recombinant Rabbit Monoclonal Antibody (HL2476)
GREENER_CHOICE

Product Code. 18076571
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18076571 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18076571 Supplier Invitrogen™ Supplier No. MA550546

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Chikungunya Virus E3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

Chikungunya virus (CHIKV) belongs to the alphavirus family, the members of which have enveloped icosahedral capsids. The maturation process of alphaviruses involves proteolysis of some of the structural proteins before assembling with nucleocapsids to produce mature virions. E3 is implicated in serine-type endopeptidase activity. Cleavage of the alphavirus precursor glycoprotein p62 into the E2 and E3 glycoproteins before assembly with the nucleocapsid is the key to producing fusion-competent mature spikes on alphaviruses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chikungunya Virus E3
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2476
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Chikungunya
Host Species Rabbit
Immunogen Full length Chikungunya virus E3 recombinant protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.