missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chikungunya Virus NSP2 Recombinant Rabbit Monoclonal Antibody (HL1432)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547042
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Chikungunya virus (CHIKV) is transmitted to humans through mosquitoes and causes Chikungunya fever Nonstructural protein 2 (nsP2) exhibits the protease and RNA helicase activities that are required for viral RNA replication and transcription [PMID: 31000599].
Specifications
| Chikungunya Virus NSP2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL1432 | |
| Unconjugated | |
| CHIKV NSP2; NsP2 protein | |
| Recombinant protein encompassing a sequence within the center region of Chikungunya virus nsP2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction