missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chikungunya Virus nsP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117443
This item is not returnable.
View return policy
Description
Centrifuge briefly prior to opening.
Chikungunya fever is a pandemic disease caused by the mosquito-borne Chikungunya virus (CHIKV). The viral enzyme non-structural protein 4 (nsP4), as RNA-dependent RNA polymerase (RdRP), catalyzes the formation of negative-sense, genomic and subgenomic viral RNAs.
Specifications
| Chikungunya Virus nsP4 | |
| Polyclonal | |
| Unconjugated | |
| Chikungunya Virus | |
| Recombinant protein encompassing a sequence within the N-terminus region of Chikungunya virus nsP4 (Chikungunya virus (strain S27-African prototype)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.27 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction