Learn More
TMC Hallcrest™ Chillchecker™ Cold Storage Indicators
Helps to identify temperature control issues during the transportation and storage of goods. TMC Hallcrest™ Chillchecker™ Cold Storage Indicators provides confirmation that the temperature has not been exceeded.
Brand:  TMC Hallcrest™ M01KTHE005C01X1
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11747218
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Confirmation that temperatures have not been exceeded
  • Easy to read and use
  • Self Adhesive
  • Available temperatures from -17 to 20°C.
  • Accurate to +/-1°C
  • Sold in units of 100
Specifications
Specifications
32 mm
Circular, Self-adhesive Temperature Indicator
41°F
41°F
41°F
5°C
PVC
1.25 in.
5°C
5°C
5°C
6 mm
Certificates
Special Offers
Special Offers
Must be stored below rated temperature