missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chinese Hamster PLBD2 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB10692-100
This item is not returnable.
View return policy
Description
PLBD2 Monoclonal antibody specifically detects PLBD2 in Chinese Hamster samples. It is validated for nan
Specifications
| PLBD2 | |
| 2751C | |
| Lyophilized from a 0.2 um filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 um filtered solution in PBS. | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 196463 | |
| Chinese Hamster | |
| Purified |
| Monoclonal | |
| Unconjugated | |
| LAMA-like protein 2,76 kDa protein, lamina ancestor homolog 2, mannose-6-phosphate protein associated protein p76, P76, phospholipase B domain containing 2, phospholipase B domain-containing protein 2, phospholipase B-like 2 32 kDa form, phospholipase B-like 2 45 kDa form, PLB homolog 2, putative phospholipase B-like 2 | |
| Recombinant Chinese Hamster PLBD-2, Met1-Asp585, Accession No. XP_003510860 | |
| 100 ug | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction