Learn More
CHIR-090, MedChemExpress
Product Code. 16486538
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16486538
Brand: Medchem Express HY15460
MedChemExpress CHIR-090 is a potent, slow, tight-binding inhibitor of the LpxC deacetylase. It binds to E. coli LpxC with a Ki of 4.0 nM.
CHIR-090 is a potent, slow, tight-binding inhibitor of the LpxC deacetylase. It binds to E. coli LpxC with a Ki of 4.0 nM.
Specifications
Chemical Name or Material CHIR-090
Physical Form Solid
CAS 728865-23-4
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H27N3O5
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 5 mg/mL (11.43 mM; Need ultrasonic)
SMILES O=C(N[C@@H]([C@@H](C)O)C(NO)=O)C(C=C1)=CC=C1C#CC2=CC=C(CN3CCOCC3)C=C2
Molecular Weight (g/mol) 437.49
Formula Weight 437.49
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only