Learn More
DiscoverX CHIR 99021 (CT99021)
Product Code. 17342449
Encompass
Click to view available options
:
2 mg
Product Code. 17342449
Brand: DiscoverX 921116
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CHIR 99021 (CT99021) is an inhibitor of GSK3α and GSK3β. Provided as 2mg of dry powder.
CHIR-99021 HCl is a synthetic small molecule and ATP-competitive inhibitor of GSK3 kinase. Has >500-fold selectivity for GSK3 over closely related CDC2 and ERK2 kinases. Functions as a WNT activator. Provided as 2mg of dry powder.
Specifications
Quantity 2 mg