Learn More
DiscoverX CHIR 99021 (CT99021)
Click to view available options
:
2 mg
Description
CHIR-99021 HCl is a synthetic small molecule and ATP-competitive inhibitor of GSK3 kinase. Has >500-fold selectivity for GSK3 over closely related CDC2 and ERK2 kinases. Functions as a WNT activator. Provided as 2mg of dry powder.
Specifications
Specifications
| Quantity | 2 mg |