missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chitinase 3-like 1 Antibody (101), Janelia Fluor™ 549 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89852JF549
This item is not returnable.
View return policy
Description
Chitinase 3-like 1 Monoclonal antibody specifically detects Chitinase 3-like 1 in Human samples. It is validated for ELISA
Specifications
| Chitinase 3-like 1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| AW208766, Brp39, Chi3l1, gp39 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Chitinase 3-like 1 (NP_001267.2; Met1-Thr383). | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 1116 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 101 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction