Learn More
Novus Biologicals™ Chitinase 3-like 1/YKL-40 Antibody (Brown U. patent anti-CHI3L1) - Chimeric, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28760-50ug
 View more versions of this product
Product Code. 30046152
  • € 213.00 / 50µg
Description
Chitinase 3-like 1/YKL-40 Monoclonal antibody specifically detects Chitinase 3-like 1/YKL-40 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chitinase 3-like 1/YKL-40
Monoclonal
Unconjugated
P36222
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Brown U. patent anti-CHI3L1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
AW208766, Brp39, Chi3l1, gp39, YKL-40
CHI3L1
50 μg
Cell Biology, Cytoskeleton Markers, Signal Transduction
1116