missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chk2 Antibody (73C175.1.1), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27403AF750
This item is not returnable.
View return policy
Description
Chk2 Monoclonal specifically detects Chk2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Chk2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CHEK2 | |
| A portion of amino acids 100-150 of human Chk2 protein was used as immunogen for this antibody. | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 73C175.1.1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CHK2 checkpoint homolog (S. pombe), EC 2.7.11, EC 2.7.11.1, S.pombe) homolog | |
| Mouse | |
| 0.1 mL | |
| 11200 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction