missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antibody detects endogenous levels of total Choline kinase alpha.
The major pathway for the biosynthesis of phosphatidylcholine occurs via the CDP-choline pathway. Choline kinase alpha is the initial enzyme in the sequence and may play a regulatory role. This protein also catalyzes the phosphorylation of ethanolamine.
Specifications
Specifications
| Antigen | CHKA |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CHKA |
| Gene Accession No. | O54804, P35790, Q01134 |
| Gene Alias | CHETK-alpha; CHK; CHKA; ChoK; Choline kinase alpha; choline kinase R; choline/ethanolamine kinase alpha; CK; CK/EK-alpha; CKI; Ckr; CK-R; EK; ethanolamine kinase; EtnK-alpha |
| Gene Symbols | CHKA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
