missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlamydia Antibody (CF 6J12), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64440B
This item is not returnable.
View return policy
Description
Chlamydia Monoclonal antibody specifically detects Chlamydia in Bacteria samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Chlamydia | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bacteria | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| CF 6J12 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Chlamydia trachomatis | |
| Elementary bodies from C. trachomatis strain SAF2. | |
| 0.5 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction