missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Chlamydia pneumoniae Monoclonal Antibody (K83A)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518183
This item is not returnable.
View return policy
Description
This antibody is specific for C. pneumoniae, with highest affinity for TWAR. It is not reactive with C. trachomatis, C. psittaci, or uninfected human cells. Purity is >90%.
Chlamydia was classified in a separate order (Chlamydiales) because of its unique developmental cycle. Chlamydia may be found in the form of an elementary body and a reticulate body and Chlamydia species have genomes of approximately 1 to 1.3 megabases in length. Chlamydia infections are the most common bacterial sexually transmitted infections in humans and are the leading cause of infectious blindness worldwide.
Specifications
| Chlamydia pneumoniae | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG2b |
| ELISA, Immunocytochemistry | |
| K83A | |
| Unconjugated | |
| C. pneumoniae | |
| Chlamydia pneumoniae native protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction