Learn More
Invitrogen™ Chlamydia trachomatis EBs Polyclonal Antibody, HRP
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA173067
Product Code. 11548441
  • € 520.00 / 1mL
Description
The PA1-73067 antibody reacts with Chlamydia trachomatis EBs. PA1-73067 has been successfully used in ELISA applications. The PA1-73067 antibody was raised against purified elementary bodies, disrupted (L2 + other serogroups).
Chlamydia trachomatis is an intracellular organism. Chlamydia usually infects the cervix and fallopian tubes of women and the urethra of men. Chlamydial infections are believed to be one of the most common of all STDs. It is generally thought that in a population of 15 million, there are up to 300,000 cases of chlamydia each year. Thus, there are many undiagnosed cases of chlamydia in the community. It has been estimated that the true prevalence of chlamydia in the sexually active population may be in the order of 5% to 10%. Chlamydia is one of the leading causes of blindness in underdeveloped countries.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chlamydia trachomatis EBs
Polyclonal
HRP
C. trachomatis; C. trachomatis EBS; Chlamydia EB; Chlamydia EBs
Purified elementary bodies, disrupted (L2 + other serogroups).
1 mL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA
1-2 mg/mL
PBS with 10mg/mL BSA and 0.002% thimerosal
Goat
IgG fraction
RUO
Bacteria