missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlamydia Trachomatis Momp Antibody (1297/143), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66403AF700
This item is not returnable.
View return policy
Description
Chlamydia Trachomatis Momp Monoclonal specifically detects Chlamydia Trachomatis Momp in Bacteria samples. It is validated for Western Blot, ELISA.
Specifications
| Chlamydia Trachomatis Momp | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| O175_03780 | |
| Native type E MOMP | |
| 0.1 mL | |
| Primary | |
| Bacteria | |
| Purified |
| Western Blot, ELISA | |
| 1297/143 | |
| Western Blot, ELISA | |
| Major Outer Membrane Protein, MOMP, omp1, omp1F, Omp1L1, ompA, ompIL2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Reacts with MOMP from C. trachomatis. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction