missing translation for 'onlineSavingsMsg'
Learn More

Chlorcyclizine HCl, MP Biomedicals

Product Code. 11456475 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456475 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11456475 Supplier MP Biomedicals Supplier No. 0520185810

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 14362-31-3
Molecular Formula C18H22Cl2N2
MDL Number MFCD00035329
Quantity 10 g
Synonym chlorcyclizine hydrochloride, chlorcyclizine hcl, histantin, eramide, perazil, di-paralene, chlorcyclizinium chloride, chlorcyclizine chloride, chlorcyclizine monohydrochloride, 1-p-chlorobenzhydryl-4-methylpiperazine monohydrochloride
InChI Key MSIJLVMSKDXAQN-UHFFFAOYNA-N
SMILES [H+].[Cl-].CN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
IUPAC Name hydrogen 1-[(4-chlorophenyl)(phenyl)methyl]-4-methylpiperazine chloride
Molecular Weight (g/mol) 337.29
PubChem CID 62413
Formula Weight 300.85
Chemical Name or Material Chlorcyclizine HCl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.