Learn More
Chlorcyclizine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Product Code. 30460389
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460389 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30460389 Supplier LoGiCal Supplier No. LGCAMP1653.0001
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Chlorcyclizine Hydrochloride
CAS 14362-31-3
Analyte or Component Names Chlorcyclizine Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C18 H21 Cl N2 . Cl H
Certifications/Compliance ISO 17034
Synonym Chlorcyclizine Hydrochloride, Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-methyl-, hydrochloride (1:1), Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-methyl-, monohydrochloride (9CI), Chlorcyclizine chloride, Chlorcyclizine hydrochloride, Chlorcyclizine monohydrochloride, Di-Paralene, Histantin, NSC 169496, Perazil, Piperazine, 1-(p-chloro-alpha-phenylbenzyl)-4-methyl-, monohydrochloride (8CI)
InChI Formula InChI=1S/C18H21ClN2.ClH/c1-20-11-13-21(14-12-20)18(15-5-3-2-4-6-15)16-7-9-17(19)10-8-16;/h2-10,18H,11-14H2,1H3;1H
Solution Type Single Solution
SMILES Cl.CN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
IUPAC Name 1-[(4-chlorophenyl)-phenylmethyl]-4-methylpiperazine;hydrochloride
Solvent or Matrix Methanol
Industry Type Food Testing
Molecular Weight (g/mol) 337.29
Quantity 1 mL
Formula Weight 336.116 g/mol