Learn More
Chlorcyclizine Hydrochloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Chlorcyclizine Hydrochloride |
| CAS | 14362-31-3 |
| Analyte or Component Names | Chlorcyclizine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H21 Cl N2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Chlorcyclizine Hydrochloride, Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-methyl-, hydrochloride (1:1), Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-methyl-, monohydrochloride (9CI), Chlorcyclizine chloride, Chlorcyclizine hydrochloride, Chlorcyclizine monohydrochloride, Di-Paralene, Histantin, NSC 169496, Perazil, Piperazine, 1-(p-chloro-alpha-phenylbenzyl)-4-methyl-, monohydrochloride (8CI) |
| InChI Formula | InChI=1S/C18H21ClN2.ClH/c1-20-11-13-21(14-12-20)18(15-5-3-2-4-6-15)16-7-9-17(19)10-8-16;/h2-10,18H,11-14H2,1H3;1H |
| Solution Type | Neat |
| SMILES | Cl.CN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3 |
| Show More |