missing translation for 'onlineSavingsMsg'
Learn More

Chlordiazepoxide EP Impurity B, Mikromol™

Product Code. 30463215
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463215 4 x 25 mg 100mg
30049365 100 mg 100mg
30050443 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30463215 Supplier Mikromol Supplier No. MM0028.0400254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Chlordiazepoxide EP Impurity B, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Chloro-2-(chloromethyl)-4-phenylquinazoline 3-Oxide
CAS 5958-24-7
Analyte or Component Names 6-Chloro-2-(chloromethyl)-4-phenylquinazoline 3-Oxide
Shipping Condition Room Temperature
Molecular Formula C15 H10 Cl2 N2 O
Certifications/Compliance ISO 17025
Synonym Chlordiazepoxide Hydrochloride Imp. B (EP), Chlordiazepoxide Imp. B (EP), 6-Chloro-2-(chloromethyl)-4-phenylquinazoline 3-Oxide, Chlordiazepoxide Hydrochloride Impurity B, Chlordiazepoxide Impurity B, Chlordiazepoxide Impurity B
InChI Formula InChI=1S/C15H10Cl2N2O/c16-9-14-18-13-7-6-11(17)8-12(13)15(19(14)20)10-4-2-1-3-5-10/h1-8H,9H2
Solution Type Neat
SMILES [O-][n+]1c(CCl)nc2ccc(Cl)cc2c1c3ccccc3
IUPAC Name 6-chloro-2-(chloromethyl)-3-oxido-4-phenylquinazolin-3-ium
Molecular Weight (g/mol) 305.16
Quantity 4 x 25 mg
Formula Weight 304.017 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.