missing translation for 'onlineSavingsMsg'
Learn More

Chlordimeform-d6, TRC

Product Code. 30362627
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362627 50 mg 50mg
30365912 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30362627 Supplier Toronto Research Chemicals Supplier No. TRCC32706250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C10 D6 H7 Cl N2
Molecular Weight (g/mol) 202.714
Synonym 1-(2-Methyl-4-chlorophenyl)-3,3-dimethylformamidine-d6, Acaron-d6, C 8514-d6, CDM-d6, CDM (acaricide)-d6, CIBA 8514-d6, Chlorophenamidin-d6, Chlorophenamidine-d6, Chlorphenamidine-d6, Fundal-d6, Fundal 500-d6, Galecron-d6, N,N-Dimethyl-N'-(2-methyl-4-chlorophenyl)formamidine-d6, N'-(2-Methyl-4-chlorophenyl)-N,N-dimethylformamidine-d6, N'-(4-Chloro-2-methylphenyl)-N,N-dimethylmethanimidamide-d6, N'-(4-Chloro-o-tolyl)-N,N-dimethylformamidine-d6, N'-(p-Chloro-o-tolyl)-N,N-dimethylformamidine-d6, RS 141-d6, Schering 36,268-d6
IUPAC Name N'-(4-chloro-2-methylphenyl)-N,N-bis(trideuteriomethyl)methanimidamide
SMILES [2H]C([2H])([2H])N(\C=N/c1ccc(Cl)cc1C)C([2H])([2H])[2H]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H13ClN2/c1-8-6-9(11)4-5-10(8)12-7-13(2)3/h4-7H,1-3H3/b12-7-/i2D3,3D3
Formula Weight 202.114
Chemical Name or Material Chlordimeform-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.