Learn More
Chlorite Standard: ClO2- @ 1000 μg/mL in H2O, VHG™
Product Code. 17909164
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 17909164
Brand: VHG Labs VHGICLO2100
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Chlorite Standard: ClO2- @ 1000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Chlorite
CAS 14998-27-7
Methods Ion Chromatography/Wet Chemistry
Analyte or Component Names Chlorite
Shipping Condition Room Temperature
Molecular Formula ClO2−
Synonym (Cl O2)-
SMILES [O-]Cl=O
IUPAC Name Chlorite
Solvent or Matrix Water
Molecular Weight (g/mol) 67.45
Quantity 100 mL
Formula Weight 67.45 g/mol