Learn More
Chlormethine hydrochloride, MedChemExpress
Product Code. 16448706
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16448706
Brand: Medchem Express HYB1253
MedChemExpress Chlormethine hydrochloride is a vesicant and necrotizing irritant destructive to mucous membranes.
Chlormethine hydrochloride is a vesicant and necrotizing irritant destructive to mucous membranes.
Specifications
Chemical Name or Material Chlormethine hydrochloride
CAS 55-86-7
Color White
Purity Grade Notes Research
Molecular Formula C5H12Cl3N
Quantity 100 mg
Synonym Mechlorethamine hydrochloride
Solubility Information DMSO : 250 mg/mL (1298.63 mM; Need ultrasonic) ∣H2O : 100 mg/mL (519.45 mM; Need ultrasonic)
SMILES ClCCN(CCCl)C.[H]Cl
Molecular Weight (g/mol) 192.51
Formula Weight 192.51
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only