missing translation for 'onlineSavingsMsg'
Learn More

Chlorodi(o-tolyl)phosphine, 98%

Product Code. 11445640
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11445640

Brand: Thermo Scientific Alfa Aesar H25890.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 36042-94-1 | C14H14ClP | 248.69 g/mol

Chlorodi(o-tolyl)phosphine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodi(o-tolyl)phosphine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
Not miscible or difficult to mix in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36042-94-1
Molecular Formula C14H14ClP
Molecular Weight (g/mol) 248.69
MDL Number MFCD04038733
InChI Key KAAGXBGJRWFWPT-UHFFFAOYSA-N
Synonym chlorodi o-tolyl phosphine, di-o-tolylchlorophosphine, bis o-tolyl chlorophosphine, chlorobis o-tolyl phosphine, chlorobis 2-methylphenyl phosphane, chloro-bis 2-methylphenyl phosphane, chloroditolylphosphine, chlorodi-o-tolylphosphine, di o-tolyl chlorophosphine, chlorobis 2-methylphenyl phosphine
PubChem CID 4590860
IUPAC Name chloro-bis(2-methylphenyl)phosphane
SMILES CC1=CC=CC=C1P(C2=CC=CC=C2C)Cl

Spécification

Melting Point 57°C
Boiling Point 174°C to 178°C (3 mmHg)
Quantity 5 g
UN Number UN1760
Sensitivity Air and moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 248.69
Percent Purity 98%
Chemical Name or Material Chlorodi(o-tolyl)phosphine

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis