missing translation for 'onlineSavingsMsg'
Learn More

Chlorodi(o-tolyl)phosphine, 98%

Product Code. p-7028525
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11435640

Brand: Thermo Scientific Alfa Aesar H25890.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 36042-94-1 | C14H14ClP | 248.69 g/mol

Chlorodi(o-tolyl)phosphine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodi(o-tolyl)phosphine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
Not miscible or difficult to mix in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36042-94-1
Molecular Formula C14H14ClP
Molecular Weight (g/mol) 248.69
MDL Number MFCD04038733
InChI Key KAAGXBGJRWFWPT-UHFFFAOYSA-N
Synonym chlorodi o-tolyl phosphine, di-o-tolylchlorophosphine, bis o-tolyl chlorophosphine, chlorobis o-tolyl phosphine, chlorobis 2-methylphenyl phosphane, chloro-bis 2-methylphenyl phosphane, chloroditolylphosphine, chlorodi-o-tolylphosphine, di o-tolyl chlorophosphine, chlorobis 2-methylphenyl phosphine
PubChem CID 4590860
IUPAC Name chloro-bis(2-methylphenyl)phosphane
SMILES CC1=CC=CC=C1P(C2=CC=CC=C2C)Cl

Specifications

Melting Point 57°C
Boiling Point 174°C to 178°C (3 mmHg)
Quantity 1 g
UN Number UN1760
Formula Weight 248.69
Percent Purity 98%
Chemical Name or Material Chlorodi(o-tolyl)phosphine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.