missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Biotech Grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer, Honeywell Burdick & Jackson™

Catalog Number p-7111953
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
67-66-3
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
trichloromethane
ClC(Cl)Cl
This item is not returnable. View return policy
67-66-3
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
trichloromethane
ClC(Cl)Cl

Biotech Grade,≥99.8%, contains 0.5-1.0% ethanol as stabilizer

  • Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
TRUSTED_SUSTAINABILITY
Melting Point -63°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Linear Formula CH3CH2OH
UN Number UN1888
Beilstein 1731042
Formula Weight 119.38
Percent Purity ≥99.8%
Grade Biotech
Density 1.48 g/mL at 25 °C; 1.492 g/mL at 25 °C(lit.)
Boiling Point 61°C
Quantity 1 L
Packaging Glass Bottle
Chemical Name or Material Chloroform, Biotech Grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'