missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloropretadalafil (Methyl (1R,3R)-1-(1,3-benzodioxol-5-yl)-2-(chloroacetyl)-2,3,4,9-tetrahydro-1H-pyrido-[3,4-b]indole-3-carboxylate), Mikromol™
Description
Chloropretadalafil (Methyl (1R,3R)-1-(1,3-benzodioxol-5-yl)-2-(chloroacetyl)-2,3,4,9-tetrahydro-1H-pyrido-[3,4-b]indole-3-carboxylate)
Specifications
Specifications
| Chemical Name or Material | Chloropretadalafil |
| CAS | 171489-59-1 |
| Analyte or Component Names | Chloropretadalafil |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C22 H19 Cl N2 O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1H-Pyrido[3,4-b]indole-3-carboxylic acid, 1-(1,3-benzodioxol-5-yl)-2-(2-chloroacetyl)-2,3,4,9-tetrahydro-, methyl ester, (1R,3R)-, 1H-Pyrido[3,4-b]indole-3-carboxylic acid, 1-(1,3-benzodioxol-5-yl)-2-(chloroacetyl)-2,3,4,9-tetrahydro-, methyl ester, (1R,3R)- (9CI), 1H-Pyrido[3,4-b]indole-3-carboxylic acid, 1-(1,3-benzodioxol-5-yl)-2-(chloroacetyl)-2,3,4,9-tetrahydro-, methyl ester, (1R-cis)-, (1R,3R)-1-(1,3-Benzodioxol-5-yl)-2-(chloroacetyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid methyl ester, Chloropretadalafil, Methyl (1R,3R)-1-(1,3-benzodioxol-5-yl)-2-(chloroacetyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate, Chloropretadanafil |
| InChI Formula | InChI=1S/C22H19ClN2O5/c1-28-22(27)16-9-14-13-4-2-3-5-15(13)24-20(14)21(25(16)19(26)10-23)12-6-7-17-18(8-12)30-11-29-17/h2-8,16,21,24H,9-11H2,1H3/t16-,21-/m1/s1 |
| SMILES | COC(=O)[C@H]1Cc2c([nH]c3ccccc23)[C@H](N1C(=O)CCl)c4ccc5OCOc5c4 |
| IUPAC Name | methyl (1R,3R)-1-(1,3-benzodioxol-5-yl)-2-(2-chloroacetyl)-1,3,4,9-tetrahydropyrido[3,4-b]indole-3-carboxylate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?