missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorpyrifos Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-88856
This item is not returnable.
View return policy
Description
Chlorpyrifos Polyclonal specifically detects Chlorpyrifos in Non-species specific samples. It is validated for ELISA, Immunoaffinity Purification.
Specifications
| Chlorpyrifos | |
| Polyclonal | |
| ELISA 1:8000, Immunoaffinity Purification | |
| chlorpyrifos-ethyl, Clorpyrifos | |
| BSA-Chlorpyrifos Conjugate | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgY |
| ELISA, Affinity Purification | |
| Unconjugated | |
| PBS | |
| Chicken | |
| PEG purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction