missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHMP2A Polyclonal Antibody
GREENER_CHOICE

Product Code. 17791174
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17791174 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17791174 Supplier Invitrogen™ Supplier No. PA5119777

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: SW480 cell lysate, A431 cell lysate, HepG2 cell lysate, rat bladder tissue. Subcellular Location: Late endosome membrane.

CHMP2A belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A is required for both MVB formation and regulation of cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP2A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene CHMP2A
Gene Accession No. O43633, Q9DB34
Gene Alias 1500016L11Rik; BC2; BC-2; Charged multivesicular body protein 2a; CHMP2; CHMP2A; chromatin modifying protein 2A; chromatin-modifying protein 2a; fj10d05; hypothetical protein LOC523847; mVps2; putative breast adenocarcinoma marker; putative breast adenocarcinoma marker (32kD); putative breast adenocarcinoma marker BC-2; RGD1305050; Vacuolar protein sorting-associated protein 2; vacuolar protein sorting-associated protein 2-1; vacuolar protein-sorting-associated protein 2; VPS2; VPS2 homolog A; vps2-1; VPS2A; wu:fi18h03; wu:fj10d05; zgc:55361
Gene Symbols CHMP2A
Host Species Rabbit
Immunogen Synthetic peptide within N-terminal human CHMP2A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27243, 365191, 68953
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.