missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHMP4B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100092
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CHMP4B.
CHMP4B is a member of the chromatin-modifying protein/charged multivesicular body protein (CHMP) protein family. The protein is part of the endosomal sorting complex required for transport (ESCRT) complex III (ESCRT-III), which functions in the sorting of endocytosed cell-surface receptors into multivesicular endosomes. The ESCRT machinery also functions in the final abscisson stage of cytokinesis and in the budding of enveloped viruses such as HIV-1. The three proteins of the CHMP4 subfamily interact with programmed cell death 6 interacting protein (PDCD6IP, also known as ALIX), which also functions in the ESCRT pathway. The CHMP4 proteins assemble into membrane-attached 5-nm filaments that form circular scaffolds and promote or stabilize outward budding. These polymers are proposed to help generate the luminal vesicles of multivesicular bodies.
Specifications
| CHMP4B | |
| Polyclonal | |
| Unconjugated | |
| CHMP4B | |
| 2010012F05Rik; C20orf178; C76846; charged multivesicular body protein 4b; CHMP4A; CHMP4B; chromatin modifying protein 4B; chromatin-modifying protein 4b; CTPP3; CTRCT31; dJ553F4.4; hSnf7-2; hVps32-2; RGD1309846; RGD1565889; Shax1; SNF7; SNF7 homolog associated with Alix 1; Snf7 homologue associated with Alix 1; Snf7-2; Vacuolar protein sorting-associated protein 32-2; vacuolar protein-sorting-associated protein 32-2; Vps32-2; VPS32B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 100359642, 128866, 75608 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9D8B3, Q9H444 | |
| CHMP4B | |
| A synthesized peptide derived from human CHMP4B(Accession Q9H444), corresponding to amino acid residues M1-E50. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?