missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP4C Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97351AF488
This item is not returnable.
View return policy
Description
CHMP4C Polyclonal antibody specifically detects CHMP4C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CHMP4C | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 92421 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| charged multivesicular body protein 4c, CHMP4c, chromatin modifying protein 4C, Chromatin-modifying protein 4c, hSnf7-3, hVps32-3, MGC22825, Shax3, SNF7 homolog associated with Alix 3, Snf7 homologue associated with Alix 3, SNF7-3, Vacuolar protein sorting-associated protein 32-3, Vps32-3, VPS32C | |
| Produced in rabbits immunized with E. coli-derived Human CHMP4C fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction