missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP5 Antibody (OTI5C7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71937B
This item is not returnable.
View return policy
Description
CHMP5 Monoclonal specifically detects CHMP5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CHMP5 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CHMP5 | |
| Full length recombinant protein of human CHMP5 (NP_057494) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5C7 | |
| Western Blot, Immunohistochemistry | |
| apoptosis-related protein PNAS-2, C9orf83, charged multivesicular body protein 5, chromatin modifying protein 5, Chromatin-modifying protein 5, chromosome 9 open reading frame 83, HSPC177, hVps60, PNAS-2, SNF7 domain containing 2, SNF7 domain-containing protein 2, SNF7DC2, Vacuolar protein sorting-associated protein 60, Vps60CGI-34 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 51510.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction