missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHMP5 Monoclonal Antibody (OTI2E6)
GREENER_CHOICE

Product Code. 15996604
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15996604 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15996604 Supplier Invitrogen™ Supplier No. MA527018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CHMP5 Monoclonal Antibody for Western Blot, IHC (P)

CHMP5 belongs to the chromatin-modifying protein/charged multivesicular body protein family. These proteins are components of ESCRT-III, a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies. Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHMP5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2E6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CHMP5
Gene Accession No. Q9NZZ3
Gene Alias 2210412K09Rik; apoptosis-related protein PNAS-2; AW545668; C9orf83; CGI-34; Charged multivesicular body protein 5; CHMP5; chromatin modifying protein 5; chromatin-modifying protein 5; HGNC:26942; HSPC177; hVps60; likely ortholog of H. sapiens SNF7 domain containing 2; PNAS-114; PNAS-2; SNF7 domain containing 2; SNF7 domain-containing protein 2; SNF7DC2; Vacuolar protein sorting-associated protein 60; Vps60
Gene Symbols CHMP5
Host Species Mouse
Immunogen Full length human recombinant protein of CHMP5 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51510
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.