missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP5 Monoclonal Antibody (OTI5F12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808525S
This item is not returnable.
View return policy
Description
CHMP5 Monoclonal antibody specifically detects CHMP5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CHMP5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13216 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1161 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5F12 | |
| Unconjugated | |
| ERCC8 | |
| 2210412K09Rik; apoptosis-related protein PNAS-2; AW545668; C9orf83; CGI-34; Charged multivesicular body protein 5; CHMP5; chromatin modifying protein 5; chromatin-modifying protein 5; HGNC:26942; HSPC177; hVps60; likely ortholog of H. sapiens SNF7 domain containing 2; PNAS-114; PNAS-2; SNF7 domain containing 2; SNF7 domain-containing protein 2; SNF7DC2; Vacuolar protein sorting-associated protein 60; Vps60 | |
| Full length human recombinant protein of human ERCC8 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction