missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16278-1-AP-20UL
This item is not returnable.
View return policy
Description
CHMP6 belongs to the chromatin-modifying protein/charged multivesicular body protein (CHMP) family. These proteins are components of ESCRT-III (endosomal sorting complex required for transport III), a complex involved in degradation of surface receptor proteins and formation of endocytic multivesicular bodies (MVBs). Some CHMPs have both nuclear and cytoplasmic/vesicular distributions, and one such CHMP, CHMP1A, is required for both MVB formation and regulation of cell cycle progression (Tsang et al., 2006 ).Specifications
| CHMP6 | |
| Polyclonal | |
| Unconjugated | |
| CHMP6 | |
| CHMP6, chromatin modifying protein 6, hVps20, VPS20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 208092, 287873, 79643 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P0C0A3, Q96FZ7 | |
| Chmp6 | |
| CHMP6 Fusion Protein Ag9357 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction