missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP7 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18468AF532
This item is not returnable.
View return policy
Description
CHMP7 Polyclonal antibody specifically detects CHMP7 in Human samples. It is validated for Western Blot
Specifications
| CHMP7 | |
| Polyclonal | |
| Western Blot | |
| charged multivesicular body protein 7, CHMP family, member 7, Chromatin-modifying protein 7, MGC29816 | |
| Recognizes region between residues Between Between 300 and 350.. | |
| 0.1 mL | |
| Autophagy, Cell Biology, Signal Transduction | |
| 91782 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction