missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHN 1 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-96984IR
This item is not returnable.
View return policy
Description
CHN 1 Polyclonal specifically detects CHN 1 in Human samples. It is validated for ELISA.
Specifications
| CHN 1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1123 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| DyLight 755 | |
| a2-chimaerin, A-chimaerin, alpha-chimerin, ARHGAP2RhoGAP2, chimerin (chimaerin) 1, CHNDuane retraction syndrome 2, DURS2, NC, N-chimaerin, N-chimerin, Rho GTPase-activating protein 2, RHOGAP2 | |
| Produced in rabbits immunized with purified, recombinant CHN 1 (NP_001813.1; Ala2-Phe459). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction