Learn More
CHN1 Mouse anti-Human, Clone: OTI2G6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505110
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GTPase-activating protein for p21-rac and a phorbol ester receptor. Involved in the assembly of neuronal locomotor circuits as a direct effector of EPHA4 in axon guidance.Specifications
| CHN1 | |
| Monoclonal | |
| Unconjugated | |
| CHN1 | |
| ARHGAP2, CHN, DURS2, NC, RHOGAP2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1123 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P15882 | |
| CHN1 | |
| Full length human recombit protein of human CHN1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.