missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cholera Toxin Beta Antibody (23043), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64675AF532
This item is not returnable.
View return policy
Description
Cholera Toxin Beta Monoclonal specifically detects Cholera Toxin Beta in Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Cholera Toxin Beta | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Mouse | |
| 0.1 mL | |
| NB100-64675 detects the labile enterotoxin normally associated with V. cholera and reacts with an epitope on the B chain. No reactivity was observed with other species tested including various E. coli serotypes. | |
| Store at 4°C in the dark. |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 23043 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
| Cholera enterotoxin B chain, Cholera enterotoxin beta chain, Cholera enterotoxin gamma chain, Cholera enterotoxin subunit B, Cholera toxin B protein, Choleragenoid, CTX B, CTXB, TOX B, toxB, VC1456 | |
| Vibrio cholerae | |
| Primary | |
| Bacteria | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction