missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cholera toxin Rabbit anti-, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB29833.500uL
This item is not returnable.
View return policy
Description
Rabbit polyclonal antibody raised against cholera toxin.
- High specificity
- High reactivity
Specifications
| Cholera toxin | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against cholera toxin. | |
| In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
| Purified choleragenoid. | |
| 500 μL | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunofluorescence, Immunohistochemistry, Lateral Flow | |
| Unconjugated | |
| ELISA Immunofluorescence Immunohistochemistry Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| IgG purification | |
| RUO | |
| This antibody reacts with beta subunit of cholera toxin. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction