Learn More
Thermo Scientific Chemicals Cholic acid, 97%
Catalog Number p-3736026
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
81-25-4
C24H40O5
MFCD00003672
BHQCQFFYRZLCQQ-OELDTZBJSA-N
cholic acid, cholalic acid, cholate, cholalin, colalin, cholsaeure, cholbam, cholicacid, cholic acid, 5beta, 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanic acid
221493
CHEBI:16359
81-25-4
C24H40O5
MFCD00003672
BHQCQFFYRZLCQQ-OELDTZBJSA-N
cholic acid, cholalic acid, cholate, cholalin, colalin, cholsaeure, cholbam, cholicacid, cholic acid, 5beta, 3alpha,7alpha,12alpha-trihydroxy-5beta-cholanic acid
221493
CHEBI:16359
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 197°C to 202°C
Color White to Beige
Loss on Drying 1% max. (135°C)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Beilstein 10, IV, 2072
Merck Index 15, 2210
Specific Rotation (20°C, 589nm) (c=2, ethanol)%gt; +35°
SMILES CC(CCC(=O)O)C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
IUPAC Name (4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molecular Weight (g/mol) 408.579
Formula Weight 408.57
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material Cholic acid