Learn More
Cholic Acid Sodium Salt, ≥98.5%, MP Biomedicals™
Product Code. p-4404522 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
100 g
1 kg
Unit Size:
100g
1kg
500g
Product Code. 11416292
Brand: MP Biomedicals 02102897.5
Cholic Acid Sodium Salt, ≥98.5% is used in the solubilization of membrane-bound proteins in their native state.
  • Cholic Acid Sodium Salt, ≥98.5% is a component for the preparation of liposomes.
  • It has also been used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.
  • Critical Micelle Concentration: 14 mM
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 361-09-1
Molecular Formula C24H39NaO5
Molecular Weight (g/mol) 430.56
MDL Number MFCD00150749,MFCD00064138
InChI Key NRHMKIHPTBHXPF-TUJRSCDTSA-M
Synonym sodium cholate, sodium 3,7,12-trihydroxycholan-24-oate, ksc575s0b, 3,7,12-trihydroxycholan-24-oic acid sodium salt
PubChem CID 57349315
IUPAC Name sodium (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate
SMILES [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
Specifications
Color White
Physical Form Powder
Quantity 5 g
Formula Weight 430.6
Percent Purity ≥98%
Chemical Name or Material Cholic acid sodium salt
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.