missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cholic Acid Sodium Salt, ≥98.5%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
100 g
1 kg
Unit Size:
100g
1kg
500g
Description
- Cholic Acid Sodium Salt, ≥98.5% is a component for the preparation of liposomes.
- It has also been used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.
- Critical Micelle Concentration: 14 mM
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 100 g |
| Formula Weight | 430.6 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Cholic acid sodium salt |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction