missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline Acetyltransferase/ChAT Antibody (8E7), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | Choline Acetyltransferase/ChAT |
|---|---|
| Clone | 8E7 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043571
|
Novus Biologicals
NBP3-26659-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043451
|
Novus Biologicals
NBP3-26659-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Choline Acetyltransferase/ChAT Monoclonal antibody specifically detects Choline Acetyltransferase/ChAT in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
| Choline Acetyltransferase/ChAT | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| acetyl CoA:choline O-acetyltransferase, ChAT, CHOACTase, Choline acetylase, choline acetyltransferase, choline O-acetyltransferase, CMS1A, CMS1A2, EC 2.3.1, EC 2.3.1.6 | |
| A synthesized peptide derived from Human Choline Acetyltransferase/ChAT [UniProt P28329] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 8E7 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 1103 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title