missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline kinase alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-85630
This item is not returnable.
View return policy
Description
Choline kinase alpha Polyclonal specifically detects Choline kinase alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Choline kinase alpha | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| CHETK-alpha, CHK, choline kinase, choline kinase alpha, CK, CKIethanolamine kinase, EC 2.7.1.32, EC 2.7.1.82, EK, Ethanolamine kinase; | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.1mg/mL | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20-1:50 | |
| P35790 | |
| CHKA | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:FDIGNHFCEWMYDYSYEKYPFFRANIRKYPTKKQQLHFISSYLPAFQNDFENLSTEEKSIIKEEMLLEVNRFALASHFLWGLWSIVQAKIS | |
| 0.1 mL | |
| Checkpoint signaling, DNA Repair | |
| 1119 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction