Learn More
Chondroitin sulfate sodium salt, Thermo Scientific Chemicals
Product Code. 15566186 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 15566186
Brand: Thermo Scientific Acros 456280010
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9082-07-9
Molecular Formula C13H21NO15S
Molecular Weight (g/mol) 463.36
InChI Key KXKPYJOVDUMHGS-OSRGNVMNSA-N
IUPAC Name (2S,3S,4S,5R,6R)-6-{[(2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-(sulfooxy)oxan-4-yl]oxy}-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CC(=O)N[C@H]1[C@H](O)O[C@H](OS(O)(=O)=O)[C@H](O)[C@@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O
Specifications
Quantity 1 g
Chemical Name or Material Chondroitin sulfate sodium salt